Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Galk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291002.1 | NP_001277931.1 | 3, 4 | 376 | ||
NM_175154.3 | NP_780363.1 | 3, 4 | 482 | |||
GenBank mRNA | AK053002.1 | - | 3, 4 | 332 | ||
AK082521.1 | - | 3, 4 | 373 | |||
AK152851.1 | - | 1, 2 | 66 | |||
AK154762.1 | - | 3, 4 | 331 | |||
AK171939.1 | - | 3, 4 | 359 | |||
BC079843.1 | - | 3, 4 | 346 | 5690056 |