Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Afap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027373.2 | NP_081649.1 | 10, 11 | 1491 | ||
GenBank mRNA | AK129483.1 | - | 10, 11 | 1418 | ||
AK147706.1 | - | 10, 11 | 1383 | |||
AK148000.1 | - | 10, 11 | 1346 | |||
AK161385.1 | - | 10, 11 | 1442 | |||
BC050814.1 | - | 10, 11 | 1491 | 6773872 |