Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Afap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027373.2 | NP_081649.1 | 11, 12 | 1629 | ||
GenBank mRNA | AK147706.1 | - | 11, 12 | 1521 | ||
AK148000.1 | - | 11, 12 | 1484 | |||
AK161385.1 | - | 11, 12 | 1580 | |||
BC050814.1 | - | 11, 12 | 1629 | 6773872 |