Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fuz | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027376.3 | NP_081652.2 | 2, 3 | 404 | ||
GenBank mRNA | AK011195.1 | - | 2, 3 | 353 | ||
AK033247.1 | - | 2, 3 | 391 | |||
AK045745.1 | - | 399 | ||||
AK134650.1 | - | 2, 3 | 404 | |||
AK168939.1 | - | 2, 3 | 399 | |||
BC045601.1 | - | 2, 3 | 390 |