Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lman1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001172062.1 | NP_001165533.1 | 1 | 182 | ||
NM_027400.3 | NP_081676.1 | 1 | 182 | |||
GenBank mRNA | AK011495.1 | - | 1 | 168 | ||
AK151599.1 | - | 1 | 169 | |||
AK151949.1 | - | 1 | 169 | |||
AK153554.1 | - | 1 | 169 | |||
AK170799.1 | - | 1 | 166 | |||
BC057165.1 | - | 1 | 191 | 4948301 | ||
BY766745.1 | - | 1 | 182 |