Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wdr77 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027432.3 | NP_081708.1 | 1 | 167 | ||
GenBank mRNA | AK012014.1 | - | 1 | 158 | ||
AK048488.1 | - | 1 | 190 | |||
AK050391.1 | - | 1 | 167 | |||
AK078856.1 | - | 2 | 126 | |||
AK161812.1 | - | 1 | 153 | |||
AK163839.1 | - | 162 | ||||
AK168006.1 | - | 1 | 169 | |||
AK168752.1 | - | 1 | 189 | |||
AK169128.1 | - | 1 | 168 | |||
BC005755.1 | - | 1 | 122 | 2654151 |