Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ckap2l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181589.3 | NP_853620.1 | 4 | 295 | ||
GenBank mRNA | AK012040.1 | - | 4 | 247 | ||
AK153766.1 | - | 4 | 254 | |||
BC053443.1 | - | 4 | 247 | 6333921 |