Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arhgap17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001122640.1 | NP_001116112.1 | 15, 16 | 1446 | ||
NM_001122641.1 | NP_001116113.1 | 15, 16 | 1446 | |||
NM_001122642.1 | NP_001116114.1 | 15, 16 | 1446 | |||
NM_001122643.1 | NP_001116115.1 | 15, 16 | 1446 | |||
NM_144529.2 | NP_653112.2 | 15, 16 | 1446 | |||
GenBank mRNA | AB060553.1 | - | 15, 16 | 1444 | ||
AB060554.1 | - | 15, 16 | 1444 | |||
AK036468.1 | - | 15, 16 | 1383 | |||
AK036617.1 | - | 15, 16 | 1383 | |||
AK046411.1 | - | 14, 15 | 1353 | |||
AK147010.1 | - | 15, 16 | 1417 | |||
AK149986.1 | - | 15, 16 | 1418 | |||
AK152266.1 | - | 15, 16 | 1383 | |||
AK152301.1 | - | 15, 16 | 1410 | |||
AK179915.1 | - | 1, 2 | 60 | |||
BC003259.1 | - | 15, 16 | 1394 | 3500822 |