Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hars2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK014515.1 | - | 7 | 811 | ||
AK077621.1 | - | 7 | 798 | |||
AK088789.1 | - | 7 | 815 | |||
AK145003.1 | - | 7 | 804 | |||
BC004596.1 | - | 7 | 762 | 3584555 |