Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nsun7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK014968.1 | - | 4 | 308 | ||
AK035181.1 | - | 5 | 823 | |||
BC115878.1 | - | 4 | 632 | 40088737 | ||
BC116658.1 | - | 4 | 632 | 40088738 |