Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spata1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027617.3 | NP_081893.1 | 4, 5 | 484 | ||
GenBank mRNA | BC028997.1 | - | 4, 5 | 517 | ||
BC042647.1 | - | 4, 5 | 517 | |||
BC055056.1 | - | 4, 5 | 545 | |||
BC139279.1 | - | 3, 4 | 397 | 8862301 | ||
BC139280.1 | - | 3, 4 | 397 | 8862302 | ||
BC171970.1 | - | 3, 4 | 397 |