Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pgm2l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027629.3 | NP_081905.1 | 9, 10 | 1543 | ||
GenBank mRNA | AK016437.1 | - | 7, 8 | 902 | ||
AK039189.1 | - | 9, 10 | 1527 | |||
AK040513.1 | - | 8, 9 | 1273 | |||
BC020351.1 | - | 9, 10 | 1756 | |||
BC032150.1 | - | 5, 6 | 749 | 5004308 | ||
BC076571.1 | - | 9, 10 | 1440 | 30356163 |