Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nipbl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027707.3 | NP_081983.2 | 33 | 6305 | ||
NM_201232.2 | NP_957684.1 | 33 | 6305 | |||
GenBank mRNA | AJ627033.1 | - | 33 | 6305 | ||
AJ640138.1 | - | 33 | 6305 | |||
AK135347.1 | - | 5 | 352 |