Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mettl13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144877.1 | NP_659126.1 | 4 | 1269 | ||
GenBank mRNA | AK050492.1 | - | 3 | 3194 | ||
AK084750.1 | - | 4 | 969 | |||
AK146751.1 | - | 1467 | ||||
AK162843.1 | - | 4 | 1454 | |||
AK168959.1 | - | 4 | 1476 | |||
AK169319.1 | - | 1468 | ||||
AK173044.1 | - | 1422 | ||||
BC014872.1 | - | 4 | 1269 | 3602469 |