Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gdpd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB048364.1 | - | 8 | 774 | ||
AK018634.1 | - | 8 | 601 | |||
AK031379.1 | - | 5 | 1557 | |||
AK086508.1 | - | 8 | 767 | |||
BC038274.1 | - | 8 | 844 | 4457335 |