Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam13c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001143776.1 | NP_001137248.1 | 13 | 1752 | ||
NM_001143777.1 | NP_001137249.1 | 14 | 1859 | |||
NM_024244.4 | NP_077206.3 | 13 | 1755 | |||
GenBank mRNA | AK004797.1 | - | 13 | 1739 | ||
AK032053.1 | - | 15 | 1594 | |||
AK041147.1 | - | 13 | 1753 | |||
AK220421.1 | - | 13 | 3297 | |||
BC003231.1 | - | 2 | 254 | |||
BC066835.1 | - | 2 | 207 | |||
BC129839.1 | - | 13 | 1708 | 40060810 | ||
BC129840.1 | - | 13 | 1711 | 40060811 |