Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrwd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027891.4 | NP_082167.2 | 9 | 1210 | ||
GenBank mRNA | AK004717.1 | - | 7 | 2172 | ||
AK051910.1 | - | 9 | 1158 | |||
AK084948.1 | - | 9 | 1210 | |||
AK153976.1 | - | 9 | 1181 | |||
AK154352.1 | - | 9 | 1152 | |||
AK167020.1 | - | 9 | 1159 | |||
AK172708.1 | - | 9 | 1178 | |||
AK218154.1 | - | 1 | 70 | |||
BC048397.1 | - | 9 | 1189 | 6396815 |