Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Slc16a14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027921.1 | NP_082197.1 | 4 | 1188 | ||
GenBank mRNA | AK003423.1 | - | 1 | 432 | ||
AK049846.1 | - | 4 | 1240 | |||
BC023456.1 | - | 4 | 1188 | 5041164 |