Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Apmap | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027977.2 | NP_082253.1 | 9 | 1277 | ||
GenBank mRNA | AJ310638.1 | - | 9 | 1296 | ||
AK009057.1 | - | 9 | 1278 | |||
AK154266.1 | - | 9 | 1302 | |||
AK205356.1 | - | 1 | 117 | |||
AK212470.1 | - | 1 | 117 | |||
BC055706.1 | - | 9 | 1278 | 6403495 |