Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Apmap | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027977.2 | NP_082253.1 | 4 | 394 | ||
GenBank mRNA | AJ310638.1 | - | 4 | 413 | ||
AK009057.1 | - | 4 | 395 | |||
AK154266.1 | - | 4 | 419 | |||
BC055706.1 | - | 4 | 395 | 6403495 |