Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zswim1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028028.2 | NP_082304.1 | 2 | 1104 | ||
GenBank mRNA | AK010358.1 | - | 2 | 778 | ||
AK149816.1 | - | 2 | 778 | |||
AK150174.1 | - | 2 | 778 | |||
BC025184.1 | - | 1 | 370 | 5040669 | ||
BC046831.1 | - | 2 | 1104 | 6812759 |