Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chst14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028117.3 | NP_082393.3 | 1 | 348 | ||
GenBank mRNA | AK011230.1 | - | 1 | 285 | ||
AK154555.1 | - | 1 | 337 | |||
AK154636.1 | - | 1 | 317 | |||
AK155115.1 | - | 1 | 331 | |||
AK159357.1 | - | 1 | 316 | |||
AK171614.1 | - | 1 | 218 | |||
BC026886.1 | - | 1 | 169 | |||
BC043700.1 | - | 1 | 321 | 5346204 | ||
BC085479.1 | - | 1 | 265 | 6415106 |