Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adpgk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028121.2 | NP_082397.1 | 7 | 1190 | ||
GenBank mRNA | AK011434.1 | - | 7 | 1155 | ||
AK149942.1 | - | 7 | 1159 | |||
AK162169.1 | - | 7 | 1144 | |||
AK167634.1 | - | 7 | 1188 | |||
AK169590.1 | - | 7 | 1154 | |||
BC021526.1 | - | 7 | 1174 | 5353637 |