Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pgm2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028132.3 | NP_082408.3 | 4, 5 | 802 | ||
GenBank mRNA | AK011485.1 | - | 4, 5 | 727 | ||
AK147982.1 | - | 4, 5 | 774 | |||
AK152678.1 | - | 4, 5 | 802 | |||
AK152923.1 | - | 4, 5 | 802 | |||
BC008527.1 | - | 4, 5 | 746 | 3495694 | ||
BC036290.1 | - | 4, 5 | 990 | |||
BC055713.1 | - | 4, 5 | 753 | |||
BC080801.1 | - | 4, 5 | 773 | |||
BC086490.1 | - | 4, 5 | 707 | 30435602 |