Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trim29 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023655.2 | NP_076144.2 | 4 | 1424 | ||
GenBank mRNA | AF230390.1 | - | 2 | 297 | ||
AK013219.1 | - | 3 | 475 | |||
AK028448.1 | - | 4 | 657 | |||
BC006699.1 | - | 4 | 1155 | |||
BC027353.1 | - | 4 | 1339 | 3988926 |