Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rusc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001083807.1 | NP_001077276.1 | 8 | 2554 | ||
NM_001083808.1 | NP_001077277.1 | 7 | 1094 | |||
NM_028188.2 | NP_082464.2 | 9 | 2965 | |||
GenBank mRNA | AK032334.1 | - | 7 | 1584 | ||
AK039664.1 | - | 9 | 2893 | |||
AK042689.1 | - | 8 | 2502 | |||
AK155528.1 | - | 1 | 2003 | |||
AK170009.1 | - | 1 | 2003 | |||
AK170012.1 | - | 7 | 1108 | |||
AK170210.1 | - | 7 | 1057 | |||
BC052081.1 | - | 6 | 1288 | |||
BC056360.1 | - | 8 | 2492 | 6816267 | ||
BC057034.1 | - | 7 | 1160 | 5687517 |