Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Htatsf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028242.2 | NP_082518.1 | 8 | 1135 | ||
NM_029371.1 | NP_083647.1 | 7 | 1059 | |||
GenBank mRNA | AK011234.1 | - | 7 | 1006 | ||
AK049495.1 | - | 7 | 991 | |||
BC037711.1 | - | 8 | 1100 | 4221465 |