Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ssbp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023672.2 | NP_076161.2 | 15, 16 | 1384 | ||
NM_198438.1 | NP_940840.1 | 14, 15 | 1303 | |||
GenBank mRNA | AF500117.1 | - | 14, 15 | 1033 | ||
AK011509.1 | - | 15, 16 | 999 | |||
AK011853.1 | - | 15, 16 | 1354 | |||
AK017711.1 | - | 15, 16 | 1388 | |||
AK041388.1 | - | 11, 12 | 900 | |||
AK133812.1 | - | 12, 13 | 842 | |||
AK153733.1 | - | 11, 12 | 744 | |||
AY167987.1 | - | 14, 15 | 916 | |||
BC003430.1 | - | 14, 15 | 1057 | 2646149 | ||
BC028324.1 | - | 9, 10 | 525 | |||
BC035475.1 | - | 14, 15 | 1246 |