Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Anks3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028301.4 | NP_082577.2 | 7 | 940 | ||
GenBank mRNA | AK012494.1 | - | 7 | 889 | ||
AK089037.1 | - | 7 | 915 | |||
AK129471.1 | - | 7 | 884 | |||
BC050929.1 | - | 7 | 876 | 6807194 | ||
BC051143.1 | - | 7 | 907 | 6531161 |