Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ints8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159595.1 | NP_001153067.1 | 8, 9 | 1160 | ||
NM_178112.5 | NP_835213.3 | 8, 9 | 1160 | |||
GenBank mRNA | AK012736.1 | - | 8, 9 | 1022 | ||
AK049513.1 | - | 8, 9 | 1131 | |||
AK140829.1 | - | 8, 9 | 1155 | |||
AK169552.1 | - | 8, 9 | 1129 | |||
BC049092.1 | - | 8, 9 | 1090 | 6402150 | ||
BC057618.1 | - | 8, 9 | 1090 | 6402150 |