Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pard3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081050.2 | NP_001074519.2 | 10, 11 | 1860 | ||
GenBank mRNA | AK008055.1 | - | 10, 11 | 1682 |