Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zdhhc4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028379.5 | NP_082655.1 | 7 | 837 | ||
GenBank mRNA | AK007572.1 | - | 5 | 823 | ||
AK013609.1 | - | 7 | 824 | |||
AK156167.1 | - | 7 | 834 | |||
BC019523.1 | - | 7 | 819 | 4163453 |