Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nme8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001167909.1 | NP_001161381.1 | 3, 4 | 234 | ||
NM_181591.3 | NP_853622.2 | 3, 4 | 234 | |||
GenBank mRNA | AF548543.1 | - | 3, 4 | 226 | ||
AK006819.1 | - | 3, 4 | 226 | |||
BC052356.1 | - | 3, 4 | 234 | 6770702 |