Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hspa12a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175199.3 | NP_780408.1 | 12 | 1907 | ||
GenBank mRNA | AK038491.1 | - | 12 | 1879 | ||
AK087732.1 | - | 13 | 2065 | |||
AK141925.1 | - | 12 | 1908 | |||
AY196789.1 | - | 12 | 1906 | |||
BC030362.1 | - | 12 | 1877 | 5369051 |