Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kif2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028547.2 | NP_082823.1 | 1 | 986 | ||
GenBank mRNA | AK030188.1 | - | 1 | 987 | ||
BC100484.1 | - | 1 | 962 | 30919380 |