Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eif3k | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001285942.1 | NP_001272871.1 | 5, 6 | 546 | ||
NM_001285943.1 | NP_001272872.1 | 5, 6 | 546 | |||
NM_028659.3 | NP_082935.1 | 6, 7 | 645 | |||
GenBank mRNA | AK004664.1 | - | 6, 7 | 572 | ||
AK158875.1 | - | 5, 6 | 523 | |||
BC027638.1 | - | 5, 6 | 445 | 1348838 | ||
BC091749.1 | - | 6, 7 | 549 | 3661337 | ||
CD802664.1 | - | 2, 3 | 218 |