Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc35b2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028662.2 | NP_082938.1 | 1 | 540 | ||
GenBank mRNA | AF414190.1 | - | 2 | 213 | ||
AK003377.1 | - | 2 | 222 | |||
AK090329.1 | - | 1 | 729 | |||
AK144403.1 | - | 2 | 236 | |||
AK152362.1 | - | 2 | 214 | |||
AK154868.1 | - | 2 | 199 | |||
AK155551.1 | - | 2 | 246 | |||
AK167343.1 | - | 1 | 541 | |||
BC025875.1 | - | 2 | 212 | 5137689 | ||
BC036992.1 | - | 2 | 220 | 5003855 |