Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ankrd42 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028665.4 | NP_082941.2 | 2 | 1075 | ||
GenBank mRNA | AK029511.1 | - | 2 | 1077 | ||
AK029888.1 | - | 2 | 1036 | |||
AK033599.1 | - | 2 | 1064 | |||
AK048925.1 | - | 2 | 969 | |||
AK133339.1 | - | 1036 | ||||
BC030885.1 | - | 2 | 704 | |||
BC100433.1 | - | 2 | 1069 | |||
BC145456.1 | - | 2 | 193 | 9054017 | ||
BC145457.1 | - | 2 | 193 | |||
BC145458.1 | - | 2 | 193 |