Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arsg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028710.3 | NP_082986.3 | 6, 7 | 1323 | ||
GenBank mRNA | AK018132.2 | - | 3, 4 | 740 | ||
AK158726.1 | - | 6, 7 | 1324 | |||
AK173082.1 | - | 3, 4 | 422 | |||
AK192752.1 | - | 2, 3 | 71 | |||
BC022158.1 | - | 2, 3 | 153 | |||
BC084731.1 | - | 3, 4 | 1013 | 6813261 |