Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sdr42e1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028725.3 | NP_083001.1 | 3 | 541 | ||
GenBank mRNA | AK014586.1 | - | 3 | 543 | ||
BC038819.1 | - | 3 | 280 | 3668862 |