Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nol9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK014568.1 | - | 8 | 1358 | ||
AK042581.1 | - | 8 | 1344 | |||
AK046761.1 | - | 8 | 1349 | |||
AK047599.1 | - | 6 | 2001 | |||
AK050948.1 | - | 8 | 1356 | |||
AK079935.1 | - | 8 | 1357 | |||
AK157398.1 | - | 8 | 1356 | |||
AK157956.1 | - | 9 | 1261 | |||
AK164493.1 | - | 8 | 1367 | |||
AK172043.1 | - | 8 | 1362 | |||
BC024877.1 | - | 8 | 1360 | |||
BC051109.1 | - | 7 | 1241 | |||
BC096599.1 | - | 7 | 1260 | |||
BC125432.1 | - | 8 | 1355 | 40129947 | ||
BC132099.1 | - | 8 | 1355 | 40130376 |