Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nebl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028757.2 | NP_083033.1 | 4, 5 | 556 | ||
GenBank mRNA | AK004645.1 | - | 4, 5 | 557 | ||
BC025863.1 | - | 4, 5 | 531 | 5249766 |