Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rbm19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028762.1 | NP_083038.1 | 20, 21 | 2521 | ||
GenBank mRNA | AK004657.1 | - | 11, 12 | 1336 | ||
AK146493.1 | - | 20, 21 | 2554 | |||
AK167100.1 | - | 20, 21 | 2518 | |||
AK216411.1 | - | 1, 2 | 68 | |||
BC025619.1 | - | 20, 21 | 2521 | 5320947 | ||
BC034010.1 | - | 20, 21 | 2487 | 4225239 |