Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Tm9sf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028780.3 | NP_083056.2 | 4, 5 | 1591 | ||
GenBank mRNA | AK004754.2 | - | 4, 5 | 1593 | ||
AK090065.1 | - | 3, 4 | 1586 | |||
AK152476.1 | - | 4, 5 | 1371 | |||
AK152540.1 | - | 4, 5 | 1371 | |||
BC007187.1 | - | 3, 4 | 896 | |||
BC017617.1 | - | 3, 4 | 1530 | 4923337 |