Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Errfi1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133753.1 | NP_598514.1 | 4 | 540 | ||
GenBank mRNA | AK004851.1 | - | 4 | 559 | ||
AK139780.1 | - | 4 | 556 | |||
AK166791.1 | - | 4 | 559 | |||
AK167156.1 | - | 4 | 499 | |||
AK167359.1 | - | 4 | 559 | |||
BC005546.1 | - | 4 | 540 | 3585640 | ||
BC057646.1 | - | 4 | 558 | 4194124 |