Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rbm26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134077.4 | NP_598838.3 | 8 | 1694 | ||
GenBank mRNA | AK049083.1 | - | 7 | 1150 | ||
AK051686.1 | - | 8 | 1670 | |||
AK139077.1 | - | 8 | 1695 | |||
AK172039.1 | - | 8 | 1673 | |||
BC066051.1 | - | 8 | 1631 |