Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tubgcp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286007.1 | NP_001272936.1 | 13 | 2287 | ||
NM_001286009.1 | NP_001272938.1 | 13 | 2287 | |||
NM_001286011.1 | NP_001272940.1 | 12 | 2054 | |||
NM_133755.2 | NP_598516.2 | 13 | 2214 | |||
GenBank mRNA | AK006233.1 | - | 3 | 318 | ||
AK033157.1 | - | 13 | 2004 | |||
AK045084.1 | - | 13 | 2003 | |||
AK146771.1 | - | 13 | 2209 | |||
AK158060.1 | - | 13 | 2003 | |||
AK167924.1 | - | 13 | 2109 | |||
BC012519.1 | - | 13 | 2142 | 3601589 | ||
BC025564.1 | - | 12 | 1976 | |||
BC025582.1 | - | 13 | 2103 | 5324981 | ||
BC027034.1 | - | 12 | 2488 |