Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wdr33 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028866.3 | NP_083142.2 | 9 | 1097 | ||
GenBank mRNA | AB086191.1 | - | 8 | 876 | ||
AK050653.1 | - | 9 | 1057 |