Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Palm3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028877.1 | NP_083153.1 | 7 | 1057 | ||
GenBank mRNA | AK014493.1 | - | 6 | 2102 | ||
BC113196.1 | - | 7 | 1060 | |||
BC113802.1 | - | 7 | 1055 | |||
EF208034.1 | - | 7 | 1057 |