Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slco6c1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028942.4 | NP_083218.1 | 4 | 948 | ||
GenBank mRNA | AK016647.1 | - | 4 | 896 | ||
AK029505.1 | - | 4 | 949 | |||
AK132668.1 | - | 4 | 899 | |||
AK161249.1 | - | 4 | 899 | |||
BC139066.1 | - | 4 | 851 | 8862088 | ||
BC139067.1 | - | 4 | 851 | 8862089 |